Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143974.1 | NP_001137446.1 | 12 - 13 | 1521 | 77 |
NM_019893.2 | NP_063946.2 | 13 - 14 | 1626 | 77 | |
XM_011539970.2 | XP_011538272.1 | 14 - 15 | 1822 | 77 | |
XM_011539971.2 | XP_011538273.1 | 14 - 15 | 1826 | 77 | |
XM_011539972.2 | XP_011538274.1 | 13 - 14 | 1646 | 77 | |
XM_011539973.2 | XP_011538275.1 | 14 - 15 | 1832 | 77 | |
XM_017016435.1 | XP_016871924.1 | 14 - 15 | 1826 | 77 | |
GenBank mRNA | AF250847.1 | - | 13 - 14 | 1569 | 77 |
AF449759.2 | - | 13 - 14 | 1626 | 77 | |
BC107105.2 | - | 12 - 13 | 1464 | 77 |