Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025010.4 | NP_079286.2 | 4 - 5 | 726 | 62 |
XM_005265001.3 | XP_005265058.2 | 4 - 5 | 626 | 62 | |
XM_005265002.4 | XP_005265059.2 | 4 - 5 | 626 | 62 | |
XM_005265003.3 | XP_005265060.2 | 4 - 5 | 626 | 62 | |
XM_006713079.1 | XP_006713142.1 | 4 - 5 | 582 | 62 | |
XM_017006033.1 | XP_016861522.1 | 3 - 4 | 489 | 62 | |
XM_017006034.1 | XP_016861523.1 | 3 - 4 | 489 | 62 | |
XM_017006035.1 | XP_016861524.1 | 3 - 4 | 489 | 62 | |
GenBank mRNA | AB018338.1 | - | 2 - 3 | 274 | 62 |
AB062478.1 | - | 4 - 5 | 898 | 62 | |
AK023765.1 | - | 4 - 5 | 621 | 62 | |
AK291477.1 | - | 4 - 5 | 620 | 62 | |
AK295292.1 | - | 3 - 4 | 488 | 62 | |
AY714879.1 | - | 1 - 2 | 153 | 62 | |
BC032620.1 | - | 4 - 5 | 538 | 62 | |
BX537953.1 | - | 4 - 5 | 617 | 62 |