Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031703.2 | NP_001026873.2 | 2 - 3 | 254 | 68 |
XM_011533851.2 | XP_011532153.1 | 2 - 3 | 256 | 68 | |
XM_011533852.2 | XP_011532154.1 | 2 - 3 | 592 | 68 | |
XM_011533853.2 | XP_011532155.1 | 2 - 3 | 233 | 68 | |
XM_017006669.1 | XP_016862158.1 | 2 - 3 | 256 | 68 | |
XM_017006670.1 | XP_016862159.1 | 2 - 3 | 525 | 68 | |
GenBank mRNA | AK000218.1 | - | 2 - 3 | 239 | 68 |
AK225721.1 | - | 2 - 3 | 236 | 68 | |
BC000623.2 | - | 2 - 3 | 158 | 68 | |
BC014204.2 | - | 2 - 3 | 158 | 68 | |
DQ787088.1 | - | 2 - 3 | 135 | 68 |