Assay Design



Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005265279.4 XP_005265336.1 28 - 29 4674 97
XM_005265280.3 XP_005265337.1 27 - 28 4491 97
XM_005265282.3 XP_005265339.1 28 - 29 4633 97
XM_005265283.3 XP_005265340.1 27 - 28 4488 97
XM_011533900.2 XP_011532202.1 30 - 31 4840 97
XM_011533902.2 XP_011532204.1 29 - 30 4772 97
XM_011533903.2 XP_011532205.1 29 - 30 4777 97
XM_017006725.1 XP_016862214.1 29 - 30 4747 97
XM_017006726.1 XP_016862215.1 30 - 31 4857 97
XM_017006727.1 XP_016862216.1 30 - 31 4802 97
XM_017006728.1 XP_016862217.1 29 - 30 4798 97
XM_017006729.1 XP_016862218.1 29 - 30 5203 97
XM_017006730.1 XP_016862219.1 30 - 31 4793 97
XM_017006731.1 XP_016862220.1 29 - 30 4776 97
XM_017006732.1 XP_016862221.1 29 - 30 4769 97
XM_017006733.1 XP_016862222.1 29 - 30 5166 97
XM_017006734.1 XP_016862223.1 29 - 30 4739 97
XM_017006735.1 XP_016862224.1 28 - 29 5160 97
XM_017006736.1 XP_016862225.1 28 - 29 5158 97
XM_017006737.1 XP_016862226.1 28 - 29 5080 97
XM_017006738.1 XP_016862227.1 28 - 29 4701 97
XM_017006739.1 XP_016862228.1 28 - 29 5077 97
XM_017006740.1 XP_016862229.1 28 - 29 4618 97
XM_017006741.1 XP_016862230.1 28 - 29 4709 97
XM_017006742.1 XP_016862231.1 28 - 29 4732 97
XM_017006743.1 XP_016862232.1 29 - 30 5131 97
XM_017006744.1 XP_016862233.1 28 - 29 4698 97
XM_017006745.1 XP_016862234.1 28 - 29 5052 97
XM_017006746.1 XP_016862235.1 28 - 29 5049 97
XM_017006747.1 XP_016862236.1 28 - 29 5111 97
XM_017006748.1 XP_016862237.1 27 - 28 5007 97
XM_017006749.1 XP_016862238.1 27 - 28 4573 97
XM_017006750.1 XP_016862239.1 26 - 27 4461 97
XM_017006751.1 XP_016862240.1 28 - 29 4634 97
XM_017006752.1 XP_016862241.1 29 - 30 5206 97
XM_017006754.1 XP_016862243.1 29 - 30 4772 97
GenBank mRNA AY281068.1 - 25 - 26 4289 97
BC129934.1 - 26 - 27 4335 97

View Full Product Details