Assay Design



Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017006725.1 XP_016862214.1 7 - 8 909 81
XM_017006726.1 XP_016862215.1 8 - 9 1019 81
XM_017006727.1 XP_016862216.1 8 - 9 964 81
XM_017006728.1 XP_016862217.1 7 - 8 960 81
XM_017006729.1 XP_016862218.1 7 - 8 1368 81
XM_017006730.1 XP_016862219.1 8 - 9 955 81
XM_017006731.1 XP_016862220.1 7 - 8 938 81
XM_017006735.1 XP_016862224.1 7 - 8 1367 81
XM_017006736.1 XP_016862225.1 7 - 8 1368 81
XM_017006737.1 XP_016862226.1 6 - 7 1242 81
XM_017006738.1 XP_016862227.1 6 - 7 863 81
XM_017006739.1 XP_016862228.1 6 - 7 1239 81
XM_017006740.1 XP_016862229.1 6 - 7 780 81
XM_017006743.1 XP_016862232.1 7 - 8 1368 81
XM_017006747.1 XP_016862236.1 7 - 8 1369 81
XM_017006752.1 XP_016862241.1 7 - 8 1368 81
XM_017006753.1 XP_016862242.1 7 - 8 1367 81

View Full Product Details