Assay Design



Gene Transcripts

Gene Symbol
PBRM1
Entrez Gene ID
55193
Gene Name
polybromo 1
Gene Aliases
BAF180, PB1
Chromosome Location
Chr.3: 52545352 - 52685933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018313.4 NP_060783.3 6 - 7 758 104
XM_005265279.4 XP_005265336.1 7 - 8 986 104
XM_005265280.3 XP_005265337.1 6 - 7 803 104
XM_005265282.3 XP_005265339.1 7 - 8 945 104
XM_005265283.3 XP_005265340.1 6 - 7 800 104
XM_011533900.2 XP_011532202.1 9 - 10 1152 104
XM_011533902.2 XP_011532204.1 8 - 9 1084 104
XM_011533903.2 XP_011532205.1 8 - 9 1089 104
XM_017006725.1 XP_016862214.1 8 - 9 1059 104
XM_017006726.1 XP_016862215.1 9 - 10 1169 104
XM_017006727.1 XP_016862216.1 9 - 10 1114 104
XM_017006728.1 XP_016862217.1 8 - 9 1110 104
XM_017006729.1 XP_016862218.1 8 - 9 1518 104
XM_017006730.1 XP_016862219.1 9 - 10 1105 104
XM_017006731.1 XP_016862220.1 8 - 9 1088 104
XM_017006732.1 XP_016862221.1 8 - 9 1084 104
XM_017006733.1 XP_016862222.1 8 - 9 1478 104
XM_017006734.1 XP_016862223.1 8 - 9 1051 104
XM_017006735.1 XP_016862224.1 8 - 9 1517 104
XM_017006736.1 XP_016862225.1 8 - 9 1518 104
XM_017006737.1 XP_016862226.1 7 - 8 1392 104
XM_017006738.1 XP_016862227.1 7 - 8 1013 104
XM_017006739.1 XP_016862228.1 7 - 8 1389 104
XM_017006740.1 XP_016862229.1 7 - 8 930 104
XM_017006741.1 XP_016862230.1 8 - 9 1066 104
XM_017006742.1 XP_016862231.1 8 - 9 1089 104
XM_017006743.1 XP_016862232.1 8 - 9 1518 104
XM_017006744.1 XP_016862233.1 8 - 9 1055 104
XM_017006745.1 XP_016862234.1 7 - 8 1364 104
XM_017006746.1 XP_016862235.1 7 - 8 1361 104
XM_017006747.1 XP_016862236.1 8 - 9 1519 104
XM_017006748.1 XP_016862237.1 7 - 8 1364 104
XM_017006749.1 XP_016862238.1 7 - 8 930 104
XM_017006750.1 XP_016862239.1 6 - 7 818 104
XM_017006751.1 XP_016862240.1 8 - 9 1066 104
XM_017006752.1 XP_016862241.1 8 - 9 1518 104
XM_017006753.1 XP_016862242.1 8 - 9 1517 104
XM_017006754.1 XP_016862243.1 8 - 9 1084 104
XM_017006755.1 XP_016862244.1 8 - 9 1066 104
XM_017006756.1 XP_016862245.1 8 - 9 1066 104
XM_017006757.1 XP_016862246.1 6 - 7 789 104
XM_017006758.1 XP_016862247.1 6 - 7 920 104
XM_017006759.1 XP_016862248.1 8 - 9 1066 104
XM_017006760.1 XP_016862249.1 8 - 9 1084 104
XM_017006761.1 XP_016862250.1 8 - 9 1093 104
XM_017006762.1 XP_016862251.1 8 - 9 1084 104
XM_017006763.1 XP_016862252.1 9 - 10 1143 104
XM_017006764.1 XP_016862253.1 8 - 9 1066 104
XM_017006765.1 XP_016862254.1 7 - 8 945 104
XM_017006766.1 XP_016862255.1 8 - 9 1073 104
GenBank mRNA AF177387.1 - 5 - 6 647 104
AF197569.1 - 6 - 7 740 104
AF225870.1 - 5 - 6 647 104
AF225871.1 - 5 - 6 647 104
AF225872.1 - 5 - 6 647 104
AK056541.1 - 4 - 5 476 104
AY281068.1 - 5 - 6 646 104
BC115009.1 - 5 - 6 662 104
BC115010.1 - 5 - 6 663 104
BC115011.1 - 5 - 6 663 104
BC129934.1 - 5 - 6 647 104
BC129935.1 - 5 - 6 647 104

View Full Product Details