Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258379.1 | NP_001245308.1 | 14 - 15 | 2224 | 135 |
NM_001258380.1 | NP_001245309.1 | 14 - 15 | 1986 | 135 | |
NM_001321103.1 | NP_001308032.1 | 15 - 16 | 2608 | 135 | |
NM_001321104.1 | NP_001308033.1 | 15 - 16 | 2569 | 135 | |
NM_001321105.1 | NP_001308034.1 | 15 - 16 | 2569 | 135 | |
NM_001321106.1 | NP_001308035.1 | 15 - 16 | 2557 | 135 | |
NM_001321107.1 | NP_001308036.1 | 14 - 15 | 2224 | 135 | |
NM_001321108.1 | NP_001308037.1 | 14 - 15 | 2236 | 135 | |
NM_003615.4 | NP_003606.3 | 15 - 16 | 2358 | 135 | |
NR_135541.1 | 14 - 15 | 2382 | 135 | ||
NR_135542.1 | 13 - 14 | 2049 | 135 | ||
NR_135543.1 | 13 - 14 | 2049 | 135 | ||
NR_135544.1 | 14 - 15 | 2382 | 135 | ||
XM_005265598.4 | XP_005265655.1 | 14 - 15 | 2490 | 135 | |
XM_005265600.4 | XP_005265657.1 | 14 - 15 | 2452 | 135 | |
XM_006713421.2 | XP_006713484.1 | 15 - 16 | 2596 | 135 | |
XM_011534256.2 | XP_011532558.1 | 15 - 16 | 2869 | 135 | |
XM_011534258.1 | XP_011532560.1 | 15 - 16 | 2572 | 135 | |
XM_011534261.2 | XP_011532563.1 | 15 - 16 | 2834 | 135 | |
XM_011534263.2 | XP_011532565.1 | 14 - 15 | 2161 | 135 | |
XM_011534265.1 | XP_011532567.1 | 14 - 15 | 2200 | 135 | |
XM_017007526.1 | XP_016863015.1 | 16 - 17 | 2493 | 135 | |
XM_017007527.1 | XP_016863016.1 | 14 - 15 | 2185 | 135 | |
XM_017007528.1 | XP_016863017.1 | 8 - 9 | 1109 | 135 | |
XM_017007529.1 | XP_016863018.1 | 8 - 9 | 1091 | 135 | |
GenBank mRNA | AB012130.1 | - | 11 - 12 | 1672 | 135 |
AF047033.1 | - | 15 - 16 | 2358 | 135 | |
AF053755.1 | - | 11 - 12 | 1664 | 135 | |
AF089726.1 | - | 14 - 15 | 1986 | 135 | |
CR627428.1 | - | 7 - 8 | 940 | 135 | |
EU499349.1 | - | 14 - 15 | 2151 | 135 | |
EU934246.1 | - | 15 - 16 | 2484 | 135 | |
EU934247.1 | - | 15 - 16 | 2496 | 135 | |
EU934248.1 | - | 15 - 16 | 2535 | 135 | |
EU934249.1 | - | 14 - 15 | 2151 | 135 | |
EU934250.1 | - | 15 - 16 | 2496 | 135 | |
FJ178574.1 | - | 14 - 15 | 2309 | 135 | |
FJ178575.1 | - | 13 - 14 | 1976 | 135 | |
FJ178576.1 | - | 13 - 14 | 1976 | 135 | |
GU354307.1 | - | 14 - 15 | 2309 | 135 | |
GU354308.1 | - | 14 - 15 | 2163 | 135 | |
GU354309.1 | - | 13 - 14 | 1988 | 135 | |
GU354310.1 | - | 13 - 14 | 1988 | 135 |