Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321103.1 | NP_001308032.1 | 1 - 2 | 355 | 77 |
NM_001321104.1 | NP_001308033.1 | 1 - 2 | 355 | 77 | |
NM_001321105.1 | NP_001308034.1 | 1 - 2 | 355 | 77 | |
NM_001321108.1 | NP_001308037.1 | 1 - 2 | 355 | 77 | |
XM_011534258.1 | XP_011532560.1 | 1 - 2 | 319 | 77 | |
XM_011534263.2 | XP_011532565.1 | 1 - 2 | 319 | 77 | |
XM_011534265.1 | XP_011532567.1 | 1 - 2 | 319 | 77 | |
GenBank mRNA | EU934247.1 | - | 1 - 2 | 282 | 77 |
EU934248.1 | - | 1 - 2 | 282 | 77 | |
EU934250.1 | - | 1 - 2 | 282 | 77 | |
GU354308.1 | - | 1 - 2 | 282 | 77 | |
GU354309.1 | - | 1 - 2 | 282 | 77 | |
GU354310.1 | - | 1 - 2 | 282 | 77 |