Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258379.1 | NP_001245308.1 | 6 - 7 | 1057 | 110 |
NM_001258380.1 | NP_001245309.1 | 6 - 7 | 819 | 110 | |
NM_001321107.1 | NP_001308036.1 | 6 - 7 | 1057 | 110 | |
NM_001321108.1 | NP_001308037.1 | 6 - 7 | 1069 | 110 | |
NR_135542.1 | 6 - 7 | 1057 | 110 | ||
NR_135543.1 | 6 - 7 | 1057 | 110 | ||
XM_005265598.4 | XP_005265655.1 | 6 - 7 | 1323 | 110 | |
XM_011534265.1 | XP_011532567.1 | 6 - 7 | 1033 | 110 | |
GenBank mRNA | AB012130.1 | - | 3 - 4 | 505 | 110 |
AF053755.1 | - | 3 - 4 | 497 | 110 | |
AF089726.1 | - | 6 - 7 | 819 | 110 | |
AK309734.1 | - | 6 - 7 | 1057 | 110 | |
EU499349.1 | - | 6 - 7 | 984 | 110 | |
EU934249.1 | - | 6 - 7 | 984 | 110 | |
FJ178575.1 | - | 6 - 7 | 984 | 110 | |
FJ178576.1 | - | 6 - 7 | 984 | 110 | |
GU354308.1 | - | 6 - 7 | 996 | 110 | |
GU354309.1 | - | 6 - 7 | 996 | 110 | |
GU354310.1 | - | 6 - 7 | 996 | 110 |