Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014249.3 | NP_055064.1 | 6 - 7 | 1194 | 62 |
NM_016346.3 | NP_057430.1 | 6 - 7 | 1194 | 62 | |
GenBank mRNA | AB307710.1 | - | 6 - 7 | 998 | 62 |
AF121129.1 | - | 6 - 7 | 1188 | 62 | |
AF148128.1 | - | 6 - 7 | 1151 | 62 | |
BC041421.1 | - | 8 - 9 | 1224 | 62 | |
HQ692847.1 | - | 6 - 7 | 998 | 62 |