Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018122.4 | NP_060592.2 | 3 - 4 | 870 | 99 |
XM_006711427.3 | XP_006711490.1 | 3 - 4 | 827 | 99 | |
XM_011509711.2 | XP_011508013.1 | 3 - 4 | 827 | 99 | |
GenBank mRNA | AK290873.1 | - | 3 - 4 | 552 | 99 |
AL833633.1 | - | 3 - 4 | 555 | 99 | |
BC045173.2 | - | 3 - 4 | 849 | 99 |