Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018122.4 | NP_060592.2 | 8 - 9 | 1332 | 99 |
XM_006711427.3 | XP_006711490.1 | 8 - 9 | 1289 | 99 | |
XM_011509711.2 | XP_011508013.1 | 8 - 9 | 1289 | 99 | |
GenBank mRNA | AK001376.1 | - | 4 - 5 | 336 | 99 |
AK022754.1 | - | 4 - 5 | 337 | 99 | |
AK290873.1 | - | 8 - 9 | 1014 | 99 | |
AL833633.1 | - | 8 - 9 | 1017 | 99 | |
BC045173.2 | - | 8 - 9 | 1311 | 99 |