Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081676.1 | NP_001075145.1 | 23 - 24 | 4584 | 84 |
NM_001081677.1 | NP_001075146.1 | 23 - 24 | 4584 | 84 | |
NM_006922.3 | NP_008853.3 | 23 - 24 | 4731 | 84 | |
XM_011511610.2 | XP_011509912.1 | 24 - 25 | 4854 | 84 | |
XM_011511613.2 | XP_011509915.1 | 23 - 24 | 4825 | 84 | |
XM_017004660.1 | XP_016860149.1 | 23 - 24 | 4714 | 84 | |
XM_017004661.1 | XP_016860150.1 | 23 - 24 | 4663 | 84 | |
XM_017004662.1 | XP_016860151.1 | 23 - 24 | 4576 | 84 | |
XM_017004663.1 | XP_016860152.1 | 24 - 25 | 4720 | 84 | |
GenBank mRNA | AF225986.1 | - | 23 - 24 | 4563 | 84 |
AF225987.1 | - | 23 - 24 | 4710 | 84 | |
AJ251507.1 | - | 23 - 24 | 4403 | 84 |