Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081676.1 | NP_001075145.1 | 8 - 9 | 1453 | 134 |
NM_001081677.1 | NP_001075146.1 | 8 - 9 | 1453 | 134 | |
NM_006922.3 | NP_008853.3 | 8 - 9 | 1453 | 134 | |
XM_011511610.2 | XP_011509912.1 | 9 - 10 | 1576 | 134 | |
XM_011511613.2 | XP_011509915.1 | 8 - 9 | 1681 | 134 | |
XM_017004660.1 | XP_016860149.1 | 8 - 9 | 1436 | 134 | |
XM_017004661.1 | XP_016860150.1 | 8 - 9 | 1436 | 134 | |
XM_017004662.1 | XP_016860151.1 | 8 - 9 | 1436 | 134 | |
XM_017004663.1 | XP_016860152.1 | 9 - 10 | 1576 | 134 | |
XM_017004664.1 | XP_016860153.1 | 8 - 9 | 1436 | 134 | |
XM_017004665.1 | XP_016860154.1 | 8 - 9 | 1436 | 134 | |
XM_017004666.1 | XP_016860155.1 | 8 - 9 | 1436 | 134 | |
GenBank mRNA | AF225986.1 | - | 8 - 9 | 1432 | 134 |
AF225987.1 | - | 8 - 9 | 1432 | 134 | |
AJ251507.1 | - | 8 - 9 | 1272 | 134 | |
AK299757.1 | - | 8 - 9 | 1436 | 134 |