Assay Design



Gene Transcripts

Gene Symbol
TGOLN2
Entrez Gene ID
10618
Gene Name
trans-golgi network protein 2
Gene Aliases
TGN38, TGN46, TGN48, TGN51, TTGN2
Chromosome Location
Chr.2: 85318018 - 85328296 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206840.1 NP_001193769.1 3 - 4 1632 68
GenBank mRNA AF027516.1 - 3 - 4 1360 68

View Full Product Details