Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014718.3 | NP_055533.2 | 5 - 6 | 1023 | 87 |
XM_006719163.3 | XP_006719226.1 | 4 - 5 | 2349 | 87 | |
GenBank mRNA | AB018269.1 | - | 4 - 5 | 1319 | 87 |
AJ277460.1 | - | 5 - 6 | 1020 | 87 | |
AK299177.1 | - | 5 - 6 | 997 | 87 | |
AK316059.1 | - | 5 - 6 | 1023 | 87 | |
AY753302.1 | - | 4 - 5 | 781 | 87 | |
BC104767.1 | - | 5 - 6 | 827 | 87 | |
BC111491.1 | - | 5 - 6 | 1011 | 87 | |
BC112283.1 | - | 5 - 6 | 839 | 87 |