Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021948.4 | NP_068767.3 | 1 - 2 | 331 | 149 |
NM_198427.1 | NP_940819.1 | 1 - 2 | 331 | 149 | |
GenBank mRNA | AK289759.1 | - | 1 - 2 | 162 | 149 |
AY358372.1 | - | 1 - 2 | 346 | 149 | |
BC009117.1 | - | 1 - 2 | 119 | 149 | |
BC010571.1 | - | 1 - 2 | 97 | 149 | |
BC022938.1 | - | 1 - 2 | 143 | 149 | |
BC027971.1 | - | 1 - 2 | 135 | 149 | |
BC035457.1 | - | 1 - 2 | 126 | 149 |