Assay Design



Gene Transcripts

Gene Symbol
BCAN
Entrez Gene ID
63827
Gene Name
brevican
Gene Aliases
BEHAB, CSPG7
Chromosome Location
Chr.1: 156641948 - 156659532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021948.4 NP_068767.3 3 - 4 802 79
NM_198427.1 NP_940819.1 3 - 4 802 79
XM_011509866.1 XP_011508168.1 3 - 4 629 79
XM_017002047.1 XP_016857536.1 3 - 4 723 79
GenBank mRNA AB209570.1 - 1 - 2 284 79
AF228710.1 - 3 - 4 513 79
AF229053.1 - 3 - 4 513 79
AK289759.1 - 3 - 4 633 79
AY358372.1 - 3 - 4 817 79
BC009117.1 - 3 - 4 590 79
BC010571.1 - 3 - 4 568 79
BC022938.1 - 3 - 4 614 79
BC027971.1 - 3 - 4 606 79
BC029313.1 - 1 - 2 86 79
BC035457.1 - 3 - 4 597 79

View Full Product Details