Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002596.3 | NP_002587.2 | 7 - 8 | 1005 | 90 |
NM_212502.2 | NP_997667.1 | 7 - 8 | 923 | 90 | |
NM_212503.2 | NP_997668.1 | 7 - 8 | 1013 | 90 | |
XM_011509602.2 | XP_011507904.1 | 7 - 8 | 1063 | 90 | |
XM_017001423.1 | XP_016856912.1 | 7 - 8 | 1158 | 90 | |
GenBank mRNA | AB209307.1 | - | 5 - 6 | 1585 | 90 |
AK126879.1 | - | 4 - 5 | 1009 | 90 | |
AK293756.1 | - | 6 - 7 | 1287 | 90 | |
AK295272.1 | - | 5 - 6 | 453 | 90 | |
AK295654.1 | - | 6 - 7 | 1068 | 90 | |
AK296320.1 | - | 7 - 8 | 971 | 90 | |
AY353237.1 | - | 7 - 8 | 838 | 90 | |
AY353238.1 | - | 7 - 8 | 902 | 90 | |
BC000281.2 | - | 1 - 2 | 90 | 90 | |
BC011526.1 | - | 7 - 8 | 858 | 90 | |
BT007299.1 | - | 6 - 7 | 655 | 90 | |
X66362.1 | - | 4 - 5 | 379 | 90 |