Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_212503.2 | NP_997668.1 | 3 - 4 | 631 | 65 |
XM_017001423.1 | XP_016856912.1 | 3 - 4 | 776 | 65 | |
GenBank mRNA | AK295272.1 | - | 1 - 2 | 71 | 65 |
AK296320.1 | - | 3 - 4 | 589 | 65 | |
AY353238.1 | - | 3 - 4 | 520 | 65 |