Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319116.1 | NP_001306045.1 | 9 - 10 | 973 | 57 |
NM_001319117.1 | NP_001306046.1 | 8 - 9 | 865 | 57 | |
NM_001319118.1 | NP_001306047.1 | 9 - 10 | 901 | 57 | |
NM_001319119.1 | NP_001306048.1 | 9 - 10 | 1181 | 57 | |
NM_001319120.1 | NP_001306049.1 | 8 - 9 | 890 | 57 | |
NM_001319121.1 | NP_001306050.1 | 10 - 11 | 874 | 57 | |
NM_001319122.1 | NP_001306051.1 | 9 - 10 | 966 | 57 | |
NM_012100.3 | NP_036232.2 | 9 - 10 | 998 | 57 | |
NR_134970.1 | 7 - 8 | 814 | 57 | ||
GenBank mRNA | AF005050.2 | - | 9 - 10 | 973 | 57 |
AK001777.1 | - | 9 - 10 | 998 | 57 | |
AK001977.1 | - | 9 - 10 | 970 | 57 | |
AK023005.1 | - | 9 - 10 | 912 | 57 | |
AK301919.1 | - | 9 - 10 | 958 | 57 | |
BC000653.2 | - | 9 - 10 | 880 | 57 | |
BC004854.2 | - | 5 - 6 | 531 | 57 |