Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170637.3 | NP_001164108.1 | 13 - 14 | 2093 | 63 |
NM_001300952.1 | NP_001287881.1 | 13 - 14 | 2093 | 63 | |
NM_015326.4 | NP_056141.2 | 13 - 14 | 2096 | 63 | |
XM_005277510.2 | XP_005277567.1 | 14 - 15 | 2283 | 63 | |
XM_005277511.2 | XP_005277568.1 | 14 - 15 | 2280 | 63 | |
XM_005277514.1 | XP_005277571.1 | 14 - 15 | 2283 | 63 | |
XM_005277515.1 | XP_005277572.1 | 14 - 15 | 2280 | 63 | |
XM_011509354.1 | XP_011507656.1 | 14 - 15 | 2283 | 63 | |
XM_011509355.1 | XP_011507657.1 | 14 - 15 | 2280 | 63 | |
XM_011509356.2 | XP_011507658.1 | 14 - 15 | 2283 | 63 | |
XM_011509359.1 | XP_011507661.1 | 14 - 15 | 2283 | 63 | |
XM_017000838.1 | XP_016856327.1 | 12 - 13 | 1927 | 63 | |
XM_017000839.1 | XP_016856328.1 | 12 - 13 | 1957 | 63 | |
XM_017000840.1 | XP_016856329.1 | 14 - 15 | 1572 | 63 | |
XM_017000841.1 | XP_016856330.1 | 14 - 15 | 2280 | 63 | |
GenBank mRNA | AB007925.1 | - | 13 - 14 | 1626 | 63 |
AK293335.1 | - | 10 - 11 | 1132 | 63 | |
AK295845.1 | - | 13 - 14 | 1849 | 63 | |
BC041635.1 | - | 5 - 6 | 397 | 63 | |
BC132872.1 | - | 13 - 14 | 1624 | 63 | |
BC132874.1 | - | 13 - 14 | 1624 | 63 | |
BC144343.1 | - | 13 - 14 | 1621 | 63 |