Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170637.3 | NP_001164108.1 | 16 - 17 | 2412 | 86 |
NM_001300952.1 | NP_001287881.1 | 16 - 17 | 2412 | 86 | |
NM_015326.4 | NP_056141.2 | 16 - 17 | 2415 | 86 | |
XM_005277510.2 | XP_005277567.1 | 17 - 18 | 2602 | 86 | |
XM_005277511.2 | XP_005277568.1 | 17 - 18 | 2599 | 86 | |
XM_005277514.1 | XP_005277571.1 | 17 - 18 | 2602 | 86 | |
XM_005277515.1 | XP_005277572.1 | 17 - 18 | 2599 | 86 | |
XM_011509354.1 | XP_011507656.1 | 17 - 18 | 2602 | 86 | |
XM_011509355.1 | XP_011507657.1 | 17 - 18 | 2599 | 86 | |
XM_011509356.2 | XP_011507658.1 | 17 - 18 | 2602 | 86 | |
XM_011509359.1 | XP_011507661.1 | 17 - 18 | 2602 | 86 | |
XM_017000838.1 | XP_016856327.1 | 15 - 16 | 2246 | 86 | |
XM_017000839.1 | XP_016856328.1 | 15 - 16 | 2276 | 86 | |
XM_017000840.1 | XP_016856329.1 | 17 - 18 | 1891 | 86 | |
XM_017000841.1 | XP_016856330.1 | 17 - 18 | 2599 | 86 | |
GenBank mRNA | AB007925.1 | - | 16 - 17 | 1945 | 86 |
AK293335.1 | - | 13 - 14 | 1451 | 86 | |
AK295845.1 | - | 16 - 17 | 2168 | 86 | |
BC041635.1 | - | 8 - 9 | 716 | 86 | |
BC132872.1 | - | 16 - 17 | 1943 | 86 | |
BC132874.1 | - | 16 - 17 | 1943 | 86 | |
BC144343.1 | - | 16 - 17 | 1940 | 86 |