Assay Design



Gene Transcripts

Gene Symbol
TTC13
Entrez Gene ID
79573
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
-
Chromosome Location
Chr.1: 230906241 - 230978872 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024525.4 NP_078801.3 17 - 18 2024 80
XM_005273261.2 XP_005273318.1 16 - 17 1931 80
XM_005273262.1 XP_005273319.1 15 - 16 1865 80
XM_005273264.2 XP_005273321.1 15 - 16 1796 80
XM_006711814.2 XP_006711877.1 13 - 14 1637 80
XM_006711815.2 XP_006711878.1 10 - 11 3031 80
XM_011544275.2 XP_011542577.1 14 - 15 1703 80
XM_011544276.2 XP_011542578.1 10 - 11 1267 80
XM_017002323.1 XP_016857812.1 13 - 14 1599 80
GenBank mRNA AK026237.1 - 4 - 5 331 80
AK074365.1 - 8 - 9 932 80
AK075386.1 - 17 - 18 1990 80
AL832286.1 - 17 - 18 1759 80

View Full Product Details