Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122835.2 | NP_001116307.2 | 7 - 8 | 867 | 132 |
NM_024525.4 | NP_078801.3 | 9 - 10 | 1026 | 132 | |
XM_005273260.1 | XP_005273317.1 | 9 - 10 | 1026 | 132 | |
XM_005273261.2 | XP_005273318.1 | 8 - 9 | 933 | 132 | |
XM_005273262.1 | XP_005273319.1 | 7 - 8 | 867 | 132 | |
XM_005273264.2 | XP_005273321.1 | 7 - 8 | 798 | 132 | |
XM_006711814.2 | XP_006711877.1 | 5 - 6 | 639 | 132 | |
XM_006711815.2 | XP_006711878.1 | 2 - 3 | 2033 | 132 | |
XM_011544275.2 | XP_011542577.1 | 6 - 7 | 705 | 132 | |
XM_011544276.2 | XP_011542578.1 | 2 - 3 | 269 | 132 | |
XM_017002321.1 | XP_016857810.1 | 8 - 9 | 933 | 132 | |
XM_017002322.1 | XP_016857811.1 | 7 - 8 | 798 | 132 | |
XM_017002323.1 | XP_016857812.1 | 5 - 6 | 601 | 132 | |
XM_017002324.1 | XP_016857813.1 | 5 - 6 | 639 | 132 | |
XM_017002325.1 | XP_016857814.1 | 9 - 10 | 4231 | 132 | |
XM_017002326.1 | XP_016857815.1 | 4 - 5 | 3611 | 132 | |
GenBank mRNA | AK075386.1 | - | 9 - 10 | 992 | 132 |
AK075561.1 | - | 7 - 8 | 846 | 132 | |
AK309725.1 | - | 5 - 6 | 685 | 132 | |
AL832286.1 | - | 9 - 10 | 761 | 132 |