Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004860.3 | NP_004851.2 | 10 - 11 | 1384 | 79 |
GenBank mRNA | AK226082.1 | - | 2 - 3 | 195 | 79 |
AK313836.1 | - | 10 - 11 | 1069 | 79 | |
BC020090.1 | - | 10 - 11 | 1338 | 79 | |
BC051907.1 | - | 10 - 11 | 1215 | 79 | |
BC067272.1 | - | 10 - 11 | 1329 | 79 | |
BT009817.1 | - | 10 - 11 | 1019 | 79 | |
U31501.1 | - | 10 - 11 | 1246 | 79 |