Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004860.3 | NP_004851.2 | 1 - 2 | 444 | 65 |
GenBank mRNA | AK313836.1 | - | 1 - 2 | 129 | 65 |
BC020090.1 | - | 1 - 2 | 398 | 65 | |
BC051907.1 | - | 1 - 2 | 275 | 65 | |
BC067272.1 | - | 1 - 2 | 389 | 65 | |
BT009817.1 | - | 1 - 2 | 79 | 65 | |
U31501.1 | - | 1 - 2 | 306 | 65 |