Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308026.1 | NP_001294955.1 | 2 - 3 | 197 | 76 |
NM_078474.2 | NP_510883.2 | 2 - 3 | 197 | 76 | |
GenBank mRNA | AK296553.1 | - | 2 - 3 | 188 | 76 |
AK315584.1 | - | 2 - 3 | 173 | 76 | |
BC006150.1 | - | 2 - 3 | 169 | 76 | |
BU595412.1 | - | 2 - 3 | 192 | 76 |