Assay Design



Gene Transcripts

Gene Symbol
DGKB
Entrez Gene ID
1607
Gene Name
diacylglycerol kinase beta
Gene Aliases
DAGK2, DGK, DGK-BETA
Chromosome Location
Chr.7: 14145049 - 14974777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004080.2 NP_004071.1 1 - 2 261 100
NM_145695.2 NP_663733.1 1 - 2 261 100
XM_005249628.2 XP_005249685.1 2 - 3 789 100
XM_005249629.2 XP_005249686.1 2 - 3 789 100
XM_005249630.2 XP_005249687.1 2 - 3 323 100
XM_005249631.3 XP_005249688.1 2 - 3 574 100
XM_011515153.2 XP_011513455.1 2 - 3 627 100
XM_011515154.2 XP_011513456.1 2 - 3 343 100
XM_011515156.2 XP_011513458.1 2 - 3 323 100
XM_011515157.2 XP_011513459.1 2 - 3 342 100
XM_011515158.2 XP_011513460.1 2 - 3 499 100
XM_017011783.1 XP_016867272.1 2 - 3 683 100
XM_017011784.1 XP_016867273.1 2 - 3 343 100
XM_017011785.1 XP_016867274.1 2 - 3 595 100
XM_017011786.1 XP_016867275.1 2 - 3 578 100
XM_017011787.1 XP_016867276.1 2 - 3 328 100
XM_017011788.1 XP_016867277.1 2 - 3 626 100
XM_017011789.1 XP_016867278.1 2 - 3 343 100
XM_017011790.1 XP_016867279.1 2 - 3 626 100
XM_017011791.1 XP_016867280.1 2 - 3 625 100
XM_017011792.1 XP_016867281.1 2 - 3 625 100
GenBank mRNA AK295668.1 - 2 - 3 595 100
AK304719.1 - 2 - 3 574 100
BC105005.1 - 1 - 2 141 100
BC143632.1 - 1 - 2 141 100
BC143633.1 - 1 - 2 141 100
DA321150.1 - 2 - 3 345 100

View Full Product Details