Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253852.2 | NP_001240781.1 | 1 - 2 | 310 | 61 |
NM_006594.4 | NP_006585.2 | 2 - 3 | 363 | 61 | |
XM_011540525.2 | XP_011538827.1 | 2 - 3 | 441 | 61 | |
XM_017000089.1 | XP_016855578.1 | 1 - 2 | 158 | 61 | |
XM_017000091.1 | XP_016855580.1 | 2 - 3 | 441 | 61 | |
XM_017000092.1 | XP_016855581.1 | 1 - 2 | 158 | 61 | |
XM_017000093.1 | XP_016855582.1 | 1 - 2 | 158 | 61 | |
GenBank mRNA | AB209817.1 | - | 1 - 2 | 247 | 61 |
AF092094.1 | - | 2 - 3 | 307 | 61 | |
AK298037.1 | - | 1 - 2 | 250 | 61 | |
AK314641.1 | - | 2 - 3 | 267 | 61 | |
BC014146.2 | - | 1 - 2 | 168 | 61 | |
DC299853.1 | - | 1 - 2 | 290 | 61 | |
DC367373.1 | - | 2 - 3 | 359 | 61 | |
DC368382.1 | - | 1 - 2 | 310 | 61 | |
DC390197.1 | - | 2 - 3 | 345 | 61 |