Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253852.2 | NP_001240781.1 | 5 - 6 | 1321 | 83 |
NM_001253853.2 | NP_001240782.1 | 6 - 7 | 1246 | 83 | |
NM_001308312.1 | NP_001295241.1 | 2 - 3 | 817 | 83 | |
NM_006594.4 | NP_006585.2 | 6 - 7 | 1374 | 83 | |
XM_011540523.2 | XP_011538825.1 | 5 - 6 | 1227 | 83 | |
XM_011540524.2 | XP_011538826.1 | 4 - 5 | 1061 | 83 | |
XM_011540525.2 | XP_011538827.1 | 4 - 5 | 1173 | 83 | |
XM_017000088.1 | XP_016855577.1 | 5 - 6 | 3058 | 83 | |
XM_017000089.1 | XP_016855578.1 | 5 - 6 | 1169 | 83 | |
XM_017000090.1 | XP_016855579.1 | 3 - 4 | 812 | 83 | |
XM_017000091.1 | XP_016855580.1 | 4 - 5 | 1173 | 83 | |
XM_017000093.1 | XP_016855582.1 | 5 - 6 | 1169 | 83 | |
GenBank mRNA | AB209817.1 | - | 5 - 6 | 1258 | 83 |
AF092094.1 | - | 6 - 7 | 1318 | 83 | |
AK001045.1 | - | 1 - 2 | 343 | 83 | |
AK093740.1 | - | 2 - 3 | 722 | 83 | |
AK300204.1 | - | 6 - 7 | 1085 | 83 | |
AK314641.1 | - | 6 - 7 | 1278 | 83 | |
BC014146.2 | - | 5 - 6 | 1179 | 83 |