Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253852.2 | NP_001240781.1 | 8 - 9 | 1714 | 66 |
NM_001253853.2 | NP_001240782.1 | 9 - 10 | 1639 | 66 | |
NM_001308312.1 | NP_001295241.1 | 5 - 6 | 1210 | 66 | |
NM_006594.4 | NP_006585.2 | 9 - 10 | 1767 | 66 | |
XM_011540523.2 | XP_011538825.1 | 8 - 9 | 1620 | 66 | |
XM_011540524.2 | XP_011538826.1 | 7 - 8 | 1454 | 66 | |
XM_011540525.2 | XP_011538827.1 | 7 - 8 | 1566 | 66 | |
XM_011540528.2 | XP_011538830.1 | 8 - 9 | 4472 | 66 | |
XM_017000088.1 | XP_016855577.1 | 8 - 9 | 3451 | 66 | |
XM_017000090.1 | XP_016855579.1 | 6 - 7 | 1205 | 66 | |
XM_017000092.1 | XP_016855581.1 | 9 - 10 | 1831 | 66 | |
GenBank mRNA | AB209817.1 | - | 8 - 9 | 1651 | 66 |
AF092094.1 | - | 9 - 10 | 1711 | 66 | |
AK057567.1 | - | 1 - 2 | 1686 | 66 | |
AK093740.1 | - | 5 - 6 | 1115 | 66 | |
AK300204.1 | - | 9 - 10 | 1478 | 66 | |
AK314641.1 | - | 9 - 10 | 1671 | 66 | |
BC014146.2 | - | 8 - 9 | 1572 | 66 |