Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287214.1 | NP_001274143.1 | 6 - 7 | 943 | 128 |
NM_001287215.1 | NP_001274144.1 | 7 - 8 | 793 | 128 | |
NM_001287216.1 | NP_001274145.1 | 3 - 4 | 670 | 128 | |
NM_001634.5 | NP_001625.2 | 7 - 8 | 1030 | 128 | |
NR_109768.1 | 4 - 5 | 713 | 128 | ||
GenBank mRNA | AJ420527.1 | - | 4 - 5 | 688 | 128 |
AK125644.1 | - | 2 - 3 | 1943 | 128 | |
AK300330.1 | - | 6 - 7 | 942 | 128 | |
AK302761.1 | - | 3 - 4 | 671 | 128 | |
AK304351.1 | - | 7 - 8 | 793 | 128 | |
AK316471.1 | - | 7 - 8 | 662 | 128 | |
AL832698.1 | - | 4 - 5 | 712 | 128 | |
BC000171.2 | - | 7 - 8 | 803 | 128 | |
BX640599.1 | - | 7 - 8 | 1030 | 128 | |
M21154.1 | - | 7 - 8 | 956 | 128 |