Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 64 - 65 | 12365 | 100 |
NM_004662.2 | NP_004653.2 | 10 - 11 | 1762 | 100 | |
XM_011523703.1 | XP_011522005.1 | 64 - 65 | 12329 | 100 | |
XM_017024292.1 | XP_016879781.1 | 63 - 64 | 12212 | 100 | |
XM_017024293.1 | XP_016879782.1 | 53 - 54 | 10273 | 100 | |
GenBank mRNA | AB002355.2 | - | 44 - 45 | 7815 | 100 |
AB290158.1 | - | 51 - 52 | 9954 | 100 | |
AF015265.1 | - | 5 - 6 | 810 | 100 | |
AF257737.1 | - | 64 - 65 | 12355 | 100 | |
AJ404468.1 | - | 64 - 65 | 12365 | 100 | |
BC128261.1 | - | 8 - 9 | 1302 | 100 | |
BC128421.1 | - | 8 - 9 | 1302 | 100 | |
BC153883.1 | - | 44 - 45 | 7815 | 100 | |
X99947.1 | - | 10 - 11 | 1762 | 100 |