Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 67 - 68 | 12915 | 77 |
NM_004662.2 | NP_004653.2 | 13 - 14 | 2312 | 77 | |
XM_011523703.1 | XP_011522005.1 | 68 - 69 | 12924 | 77 | |
XM_017024292.1 | XP_016879781.1 | 67 - 68 | 12807 | 77 | |
XM_017024293.1 | XP_016879782.1 | 57 - 58 | 10868 | 77 | |
GenBank mRNA | AB002355.2 | - | 47 - 48 | 8365 | 77 |
AB290158.1 | - | 54 - 55 | 10504 | 77 | |
AF257737.1 | - | 67 - 68 | 12905 | 77 | |
AJ404468.1 | - | 67 - 68 | 12915 | 77 | |
BC128261.1 | - | 11 - 12 | 1852 | 77 | |
BC128421.1 | - | 11 - 12 | 1852 | 77 | |
BC153883.1 | - | 47 - 48 | 8365 | 77 | |
X99947.1 | - | 13 - 14 | 2312 | 77 |