Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001372.3 | NP_001363.2 | 2 - 3 | 674 | 97 |
XM_011523703.1 | XP_011522005.1 | 2 - 3 | 638 | 97 | |
XM_017024292.1 | XP_016879781.1 | 2 - 3 | 638 | 97 | |
XM_017024294.1 | XP_016879783.1 | 2 - 3 | 638 | 97 | |
XM_017024295.1 | XP_016879784.1 | 2 - 3 | 638 | 97 | |
GenBank mRNA | AF257737.1 | - | 2 - 3 | 664 | 97 |
AJ404468.1 | - | 2 - 3 | 674 | 97 | |
AK055990.1 | - | 2 - 3 | 636 | 97 | |
AK310137.1 | - | 2 - 3 | 633 | 97 |