Assay Design



Gene Transcripts

Gene Symbol
DNAH9
Entrez Gene ID
1770
Gene Name
dynein axonemal heavy chain 9
Gene Aliases
DNAH17L, DNEL1, DYH9, Dnahc9, HL-20, HL20
Chromosome Location
Chr.17: 11598431 - 11970168 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372.3 NP_001363.2 53 - 54 10553 116
XM_011523703.1 XP_011522005.1 53 - 54 10517 116
XM_017024292.1 XP_016879781.1 52 - 53 10400 116
XM_017024293.1 XP_016879782.1 42 - 43 8461 116
GenBank mRNA AB002355.2 - 34 - 35 6231 116
AB290158.1 - 41 - 42 8370 116
AF257737.1 - 53 - 54 10543 116
AJ404468.1 - 53 - 54 10553 116
BC153883.1 - 34 - 35 6231 116

View Full Product Details