Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001622.2 | NP_001613.2 | 5 - 6 | 750 | 64 |
GenBank mRNA | AK292751.1 | - | 5 - 6 | 747 | 64 |
AK303754.1 | - | 5 - 6 | 887 | 64 | |
AK312969.1 | - | 5 - 6 | 749 | 64 | |
BC048198.1 | - | 5 - 6 | 731 | 64 | |
BC052590.1 | - | 5 - 6 | 730 | 64 | |
D67012.1 | - | 5 - 6 | 570 | 64 | |
M16961.1 | - | 5 - 6 | 717 | 64 |