Assay Design
Gene Transcripts
- Gene Symbol
- ALKBH6
- Entrez Gene ID
-
84964
- Gene Name
- alkB homolog 6
- Gene Aliases
-
ABH6
- Chromosome Location
-
Chr.19: 36009116 - 36014243 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK310704.1 |
- |
4 - 5 |
273 |
115 |
BC007601.1 |
- |
4 - 5 |
245 |
115 |
View Full Product Details