Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014396.3 | NP_055211.2 | 24 - 25 | 2146 | 92 |
NM_080631.3 | NP_542198.2 | 23 - 24 | 2071 | 92 | |
XM_017011988.1 | XP_016867477.1 | 11 - 12 | 1079 | 92 | |
GenBank mRNA | AK026848.1 | - | 3 - 4 | 249 | 92 |
AK074325.1 | - | 20 - 21 | 1859 | 92 | |
AK299143.1 | - | 21 - 22 | 1892 | 92 | |
AK314556.1 | - | 24 - 25 | 2146 | 92 | |
BC044851.1 | - | 24 - 25 | 2141 | 92 | |
BX648347.1 | - | 22 - 23 | 1871 | 92 | |
L40398.1 | - | 3 - 4 | 279 | 92 | |
U87281.1 | - | 2 - 3 | 161 | 92 | |
U87309.1 | - | 24 - 25 | 2149 | 92 |