Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014396.3 | NP_055211.2 | 8 - 9 | 594 | 84 |
NM_080631.3 | NP_542198.2 | 7 - 8 | 519 | 84 | |
GenBank mRNA | AK074325.1 | - | 4 - 5 | 307 | 84 |
AK299143.1 | - | 5 - 6 | 340 | 84 | |
AK314556.1 | - | 8 - 9 | 594 | 84 | |
BC044851.1 | - | 8 - 9 | 589 | 84 | |
U87309.1 | - | 8 - 9 | 597 | 84 |