Assay Design



Gene Transcripts

Gene Symbol
CLEC4M
Entrez Gene ID
10332
Gene Name
C-type lectin domain family 4 member M
Gene Aliases
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
Chromosome Location
Chr.19: 7763052 - 7770559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006722612.3 XP_006722675.1 2 - 3 338 72
GenBank mRNA AK298084.1 - 2 - 3 153 72

View Full Product Details