Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014445.2 | NP_001014445.1 | 1 - 2 | 275 | 64 |
NM_018096.4 | NP_060566.2 | 2 - 3 | 204 | 64 | |
XM_017024777.1 | XP_016880266.1 | 2 - 3 | 193 | 64 | |
GenBank mRNA | AB209111.1 | - | 1 - 2 | 275 | 64 |
AK001320.1 | - | 2 - 3 | 193 | 64 | |
AK303253.1 | - | 2 - 3 | 179 | 64 | |
AK304508.1 | - | 2 - 3 | 192 | 64 | |
BC002884.2 | - | 1 - 2 | 162 | 64 | |
BC012075.1 | - | 2 - 3 | 177 | 64 | |
BI752930.1 | - | 1 - 2 | 297 | 64 | |
BI911995.1 | - | 2 - 3 | 185 | 64 | |
BP244661.1 | - | 2 - 3 | 64 |