Assay Design



Gene Transcripts

Gene Symbol
NLE1
Entrez Gene ID
54475
Gene Name
notchless homolog 1
Gene Aliases
Nle
Chromosome Location
Chr.17: 35128755 - 35142315 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014445.2 NP_001014445.1 4 - 5 647 61
NM_018096.4 NP_060566.2 5 - 6 576 61
XM_017024777.1 XP_016880266.1 5 - 6 594 61
GenBank mRNA AB209111.1 - 4 - 5 647 61
AK001320.1 - 5 - 6 565 61
BC002884.2 - 4 - 5 534 61
BC012075.1 - 5 - 6 549 61
BE272051.1 - 1 - 2 52 61
BI911995.1 - 5 - 6 557 61
BQ070427.1 - 3 - 4 308 61

View Full Product Details