Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031898.2 | NP_114104.1 | 7 - 8 | 1295 | 102 |
XM_011523988.2 | XP_011522290.1 | 7 - 8 | 1190 | 102 | |
XM_011523989.1 | XP_011522291.1 | 6 - 7 | 1252 | 102 | |
XM_011523990.2 | XP_011522292.1 | 8 - 9 | 2612 | 102 | |
XM_017024954.1 | XP_016880443.1 | 7 - 8 | 1339 | 102 | |
XM_017024955.1 | XP_016880444.1 | 7 - 8 | 1035 | 102 | |
GenBank mRNA | AF334676.1 | - | 6 - 7 | 1207 | 102 |
AK057390.1 | - | 7 - 8 | 1295 | 102 | |
AK223534.1 | - | 7 - 8 | 1251 | 102 | |
AK314327.1 | - | 7 - 8 | 1287 | 102 | |
BC031688.1 | - | 7 - 8 | 1258 | 102 |