Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020777.2 | NP_065828.2 | 23 - 24 | 3110 | 85 |
XM_005247987.4 | XP_005248044.2 | 23 - 24 | 3150 | 85 | |
XM_011513514.2 | XP_011511816.1 | 23 - 24 | 3150 | 85 | |
XM_011513515.2 | XP_011511817.1 | 23 - 24 | 3150 | 85 | |
XM_011513516.2 | XP_011511818.1 | 23 - 24 | 3150 | 85 | |
XM_017008481.1 | XP_016863970.1 | 23 - 24 | 3150 | 85 | |
GenBank mRNA | AB037750.1 | - | 20 - 21 | 2354 | 85 |
AK295629.1 | - | 23 - 24 | 2726 | 85 |