Assay Design



Gene Transcripts

Gene Symbol
SORCS2
Entrez Gene ID
57537
Gene Name
sortilin related VPS10 domain containing receptor 2
Gene Aliases
-
Chromosome Location
Chr.4: 7192607 - 7742837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020777.2 NP_065828.2 25 - 26 3311 68
XM_011513514.2 XP_011511816.1 25 - 26 3351 68
GenBank mRNA AB037750.1 - 22 - 23 2555 68

View Full Product Details