Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004169.4 | NP_004160.3 | 4 - 5 | 566 | 92 |
NM_148918.2 | NP_683718.1 | 4 - 5 | 566 | 92 | |
XM_005256767.3 | XP_005256824.1 | 5 - 6 | 637 | 92 | |
XM_011523992.2 | XP_011522294.1 | 4 - 5 | 534 | 92 | |
XM_017024957.1 | XP_016880446.1 | 4 - 5 | 391 | 92 | |
XM_017024958.1 | XP_016880447.1 | 5 - 6 | 637 | 92 | |
GenBank mRNA | AK223552.1 | - | 4 - 5 | 497 | 92 |
AK302834.1 | - | 4 - 5 | 504 | 92 | |
BC007979.2 | - | 4 - 5 | 412 | 92 | |
BC022874.1 | - | 4 - 5 | 416 | 92 | |
BC038598.1 | - | 4 - 5 | 423 | 92 | |
L11931.1 | - | 3 - 4 | 379 | 92 | |
L23928.1 | - | 3 - 4 | 367 | 92 | |
Y14485.1 | - | 4 - 5 | 532 | 92 |