Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122835.2 | NP_001116307.2 | 15 - 16 | 1858 | 77 |
XM_005273260.1 | XP_005273317.1 | 17 - 18 | 2017 | 77 | |
XM_017002321.1 | XP_016857810.1 | 16 - 17 | 1924 | 77 | |
XM_017002322.1 | XP_016857811.1 | 15 - 16 | 1789 | 77 | |
XM_017002324.1 | XP_016857813.1 | 13 - 14 | 1630 | 77 | |
XM_017002325.1 | XP_016857814.1 | 17 - 18 | 5222 | 77 | |
XM_017002326.1 | XP_016857815.1 | 12 - 13 | 4602 | 77 | |
GenBank mRNA | AK075561.1 | - | 15 - 16 | 1837 | 77 |
AK309725.1 | - | 13 - 14 | 1676 | 77 | |
AL834453.1 | - | 8 - 9 | 986 | 77 | |
BC042683.1 | - | 8 - 9 | 904 | 77 |