Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000240.3 | NP_000231.1 | 13 - 14 | 2803 | 60 |
NM_001270458.1 | NP_001257387.1 | 14 - 15 | 2911 | 60 | |
GenBank mRNA | AK223499.1 | - | 13 - 14 | 1545 | 60 |
AK291769.1 | - | 13 - 14 | 1407 | 60 | |
AK293926.1 | - | 14 - 15 | 1518 | 60 | |
BC008064.2 | - | 13 - 14 | 1389 | 60 | |
BC044787.1 | - | 13 - 14 | 2805 | 60 | |
BT006651.1 | - | 13 - 14 | 1368 | 60 | |
M68840.1 | - | 13 - 14 | 1441 | 60 | |
M69226.1 | - | 13 - 14 | 1418 | 60 |